PYREX® Hollow [ST] Flask Stoppers, Corning
Supplier: Corning
Certificates
About this item
These PYREX® hollow borosilicate glass stoppers are light weight, strong to reduce chipping and breakage. Barrel shaped head makes it easy to clean and rotate in or out. Grooves in the sides help prevent slippage.
- Interchangeable with standard taper joints
Specifications
- Catalog No:
- 59432-298
- 59432-276
- 59432-243
- 59432-210
- 59432-312
- 59432-185
- 59432-152
- 59432-120
- 59432-108
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey