Nunc® Lab-Tek™ Chambered Coverglasses, Sterile, Thermo Scientific
Supplier: Avantor
Certificates
About this item
This cell culture vessel is mounted onto a No.1 borosilicate coverglass
The non-removable glass is thin to allow examination using inverted microscope systems at high magnification levels. The polystyrene upper structure is attached to the glass with a non-toxic medical grade silicone adhesive.
Ordering information: Supplied sterile, 8 per incubation tray.
Related Information
Certificates
Search by Lot NumberThermo Scientific Nunc Cell Culture Plastics Guide
Product Literature
Customers Who Bought This Also Bought