SPILFYTER® Chemical Spill Center, NPS
Catalog #: 56609-224
Supplier: NPS
Specifications
- Description:Chemical Spill Center
- Cat. no.:56609-224
- Supplier No.:200800
Specifications
About this item
Keep response materials close at hand in spill-prone areas with this wall-mounted system.
- Space-saving cabinet contains emergency response products and protective gear
- Provides quick response capability for liquid spills
- Cabinet measures 155/8 Wx181/2 Hx91/8 D"
Ordering information: Spill center contents include: 10 universal sorbent pillows (81/2x17"); 10 universal sorbent sheets (8x12"); 2 pair nitrile gloves; 2 poly aprons; 2 pair chemical splash goggles and 10 disposal bags.
Customers Who Bought This Also Bought