VWR® Corks
Catalog #: CA23417-000
Supplier: VWR International
Specifications
- Description:Assorted, Nos. 12 to 26
- Environmentally Preferable:Low Manufacturing Impact
- Cat. no.:CA23417-000
Specifications
About this item
XXXX Corks function as a closure for bottles, vials, laboratory and other openings.
- Regular length taper
- Assorted sizes
The XXXX premium grade corks have fewer lenticels (crevices) than standard grade. This means they are better suited for use as laboratory corks and for transporting many kinds of liquids.
Customers Who Bought This Also Bought