T-Bore Stopcocks, with Glass Plug, Chemglass
Supplier: Chemglass
Certificates
About this item
Stopcocks have three sidearms sealed at 90° angles
The [ST] solid glass plugs have a T-shaped bore to allow any or all of the three sidearms to be connected at one time.
Ordering information: All stopcocks are supplied with a metal retaining clip.
Customers Who Bought This Also Bought