Tygon® Fuel and Lubricant Tubing, Formulation F-4040-A, Saint-Gobain Life Sciences
Supplier: Saint Gobain Life Sciences
Certificates
About this item
Tygon® F-4040-A tubing is used for petroleum and petroleum/alcohol blends.
Flexible and resistant to swelling and brittleness. Minimum extractability safeguards fluids being handled. Translucent yellow color for positive identification and easy flow monitoring.
Recommended operating temperature: –37 °C (–35 °F) to 74 °C (165 °F). Durometer hardness: Shore A, 57. Tensile strength: 1821 psi. Not for use with strong acids or alkalies.
Suitable for transporting gasoline, heating oils, kerosene, and glycol-based coolants.
Ordering information: Available in 50' coil lengths.
Related Information
Certificates
Search by Lot Number63010-406_2454858 63010-428_23843828 63010-450_31585705 63010-461_24452343
Conformity declaration
Customers Who Bought This Also Bought