TMCS Silylation Reagent
Catalog #: 10066-408
Supplier: Thermo Scientific
CAS Number:
Specifications
- Description:TMCS silylation reagent
- Size:25 g
- Temperature Limit:Room temperature
- Supplier No.:TS-88530
Specifications
About this item
Catalyze the derivatization of difficult-to-silylate compounds using Thermo Scientific™ TMCS silylation reagent.
- Excellent adjunct for forming trimethylsilyl ethers for GC determinations
- Used to prepare TMS derivatives of organic acids
Standard concentration: 98 to 100%
Hazards: Flammable, toxic
Standard components: Chlorotrimethylsilane
Customers Who Bought This Also Bought