Long Neck Spherical Dewar Flasks, Pope Scientific
Catalog #: 63405-109
Supplier: Pope Scientific
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- Capacity:2 L
- Description:Dewar Flask
- Supplier No.:8697
Specifications
About this item
Spherical Dewar flasks
Made with pads between the borosilicate glass walls. Designed to give the greatest efficiency in holding liquid nitrogen (LN2). Ideal for use with liquid gases.
Customers Who Bought This Also Bought