SpectroBlend® additive, powder, Chemplex® 44 µm
Catalog #: 15566-078
Supplier: Chemplex
CAS Number:
Specifications
- Size:454 g
- Cat. no.:15566-078
- Supplier No.:660
Specifications
Specification Test Results
- SpectroBlend® additive, powder, Chemplex® 44 µm:
Specification Test Results
About this item
:Length:
:Width:
:Height:
:Ø
:Thickness:
:44 µm
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
Customers Who Bought This Also Bought