BradyGlo® Frameless Exit Signs, Brady®
Supplier: Brady Worldwide
Certificates
About this item
Exit signs are printed on photoluminescent plastic and supplied with pressure-sensitive directional indicators to be applied by the end user
Signs are NFPA and OSHA compliant. UL and C-UL listed.
Glow-in-the-dark signs require no electrical wiring or batteries. White background. Approved for use with fluorescent, mercury vapor, and metal halide light sources.
Customers Who Bought This Also Bought