To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Protein Stabilizing Cocktail
- Size:10 mL
- Cat. no.:CAPI89806
- Supplier No.:89806
Specifications
About this item
A versatile stabilizing solution that increases the shelf-life of purified or partially purified proteins during routine storage.
- Stabilizes most enzymes and other proteins significantly better than ordinary buffers
- Protect proteins from environmental stresses
- Does not destabilize biomolecules in downstream assays
- Components are low molecular weight and fully dialyzable
- Low-viscosity reagent is easier to pipette than 50 % glycerol
The formulation is comprised of low-weight, naturally occurring molecules that protect from environmental stresses that could lead to enzyme inactivation, aggregation, and freeze-thaw damage
Protein classes tested include kinases, phosphatases, peroxidases, restriction enzymes, luciferases, cytokines and antibodies.
In addition, it will not destabilize biomolecules in downstream assays. However, if needed, all components of the reagent can be removed by dialysis or desalting before the sample proteins are used in downstream assays. The solution is a 4X concentrate.