Write-On Hazard Labels, National Marker
Catalog #: 56606-751
Supplier: ACCUFORM MANUFACTURING, INC
Specifications
- Description:Write-on hazard labels
- Cat. no.:56606-751
- Dimensions:101.6×102 mm (4×4")
- Supplier No.:HMS14L
Specifications
About this item
Paper write-on NFPA hazard label identifies the degree of health hazard, flammability, reactivity and specific hazards of chemicals.
Colored squares can be numbered to the NFPA rating system of 0 to 4. White square is used for specific hazard warning.
Customers Who Bought This Also Bought