Specifications
- Size:1 set of 2×1 ml
- Enzyme name:Peroxynitrite
- Cat. no.:CA80055-374
- Supplier No.:516620-1SET
Specifications
About this item
Synthesized from isoamyl nitrite and hydrogen peroxide
Peroxynitrite (ONOO-) is a cell-permeable strong biological oxidizing agent that reacts with DNA, membrane phospholipids, sulfhydryl groups, and tyrosine. Has been implicated in a variety of pathophysiological conditions including acute respiratory distress syndrome (ARDS), arthritis, atherosclerosis, inflammatory bowel disease, ischemia-reperfusion, N-methyl-4-phenyl-1,2,3,6-tetrahydropyridine (MPTP)-mediated neurotoxicity, and septic shock. Supplied with an informational insert.