Uvex Stealth® Goggles, Honeywell Safety
Supplier: Honeywell Safety Products
Certificates
About this item
Protect against dust, mist, and splashes with these stylish goggles.
- Durable polypropylene frame and soft, flexible elastomer provide comfortable seal
- Indirect ventilation reduces fogging
- Uvextreme® lenses provide antifog, anti-fog, anti-scratch, anti-static
- Dura-streme® lenses feature interior Uvextreme® coating and exterior Ultra-dura® coating for enhanced durability
- One-piece, impact-resistant polycarbonate lens can be replaced quickly and easily
Lenses are available with either the exclusive Uvextreme® AF or Dura-streme™ lens coatings. Dura-streme™ lens coating features a combination of Ultra-dura® on the exterior and Uvextreme® on the interior for excellent protection and longevity. The Rx Carrier goggle insert is an accessory item designed to hold prescription lenses in place inside the goggle.
Durable polypropylene frame, combined with soft, flexible elastomer, conforms to facial contours for a comfortable seal.
Certifications: Meet ANSI Z87+ and CSA Z94.3 high impact protector standards.