N-Acetyl-L-cysteine ≥98%, Calbiochem®
Catalog #: CA80057-756
Supplier: MilliporeSigma
CAS Number: 616-91-1
Specifications
- Packaging:Bottle
- Size:5 g
- Cat. no.:CA80057-756
- Supplier No.:106425-5GM
Specifications
About this item
Synonyms: Acetylcysteine,(R)-(+)-N-Acetylcysteine,(R)-2-Acetamido-3-mercaptopropanoic acid
Boiling Pt:371 °C (1013 hPa)
Melting Pt:106…108 °C
Density:1.48 g/cm³ (20 °C)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:98
:Boiling Pt:
:Boiling Pt: 371 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 106…108 °C
:Flash Pt:
:Flash Pt:
:M.W. 163.2 g/mol
Merck Index:13,00090
Formula:C₅H₉NO₃S
MW:163.2 g/mol
Storage Temperature:Ambient
MDL Number:MFCD00004880
CAS Number:616-91-1
EINECS:210-498-3
Customers Who Bought This Also Bought