Specifications
- Size:25 mg
- Enzyme name:FeTPPS
- Cat. no.:CA80056-448
- Supplier No.:341492-25MG
Specifications
About this item
A ferric porphyrin complex that catalytically isomerizes peroxynitrite to nitrate both in vivo and in vitro.
Provides cytoprotection against peroxynitrite (EC50=5 µM). Does not complex with nitric oxide and exhibits minimal SOD mimetic activity.