Specifications
- Size:25 mg
- Enzyme name:MnTBAP
- Cat. no.:CA80053-226
- Supplier No.:475870-25MG
Specifications
About this item
Cell-permeable superoxide dismutase (SOD) mimetic and peroxynitrite scavenger. Protects endothelial cells in a dose-dependent manner (EC50~40 µM) against paraquat (2 mM). Inhibits the oxidation of Dihydrorhodamine 123 by peroxynitrite, but does not scavenge nitric oxide (NO).