Coelenterazine ≥90% (by TLC), Calbiochem®, Sigma-Aldrich®
Catalog #: CA80051-104
Supplier: MilliporeSigma
CAS Number: 55779-48-1
Specifications
- Packaging:Plastic ampoule
- Size:250 μg
- Cat. no.:CA80051-104
- Supplier No.:233900-250UG
Specifications
Specification Test Results
- Coelenterazine ≥90%(by TLC), Calbiochem®, Sigma-Aldrich®:
Specification Test Results
About this item
Synonyms: 8-Benzyl-2-(4-hydroxybenzyl)-6-(4-hydroxyphenyl)imidazo[1,2-a]pyrazin-3(7H)-one,CLZN,2-[(4-Hydroxyphenyl)methyl]-6-(4-hydroxyphenyl)-8-(phenylmethyl)-imidazo [1
:Length:
:Width:
:Height:
:Ø
:Thickness:
:90
:M-Clarity™ quality level = MQ100
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 423.472 g/mol
:Coelenterazine
MW:423.472 g/mol
Storage Temperature:Freezer
:MQ100
Formula:C₂₆H₂₁N₃O₃
MDL Number:MFCD00467176
CAS Number:55779-48-1
Customers Who Bought This Also Bought