Crews® CK2® Protective Eyewear, MCR Safety
Supplier: Avantor
Certificates
About this item
These super-lightweight glasses provide a nearly complete orbital shield around the eyes for maximum protection.
- Polycarbonate single-lens glasses with wraparound side shields are scratch-resistant
- Anti-fog lenses feature Duramass AF4® antifog lens coating
Lenses filter out 99% of UV radiation. Soft, secure gel nosepiece and gel temple sleeves provide a comfortable fit for extended wear.
Certifications: Meets or exceeds ANSI Z87+ standards.
Customers Who Bought This Also Bought