About this item
The TurboBlotter™ system is a rapid, downward blotting device for high-resolution transfer of DNA and RNA. The conventional Northern/Southern transfer stack has been turned upside down in order to take advantage of gravity.
- Rapid - downward capillary transfer allows for alkaline buffer transfers in one hour and neutral (SSC) transfers in three hours
- Economical - reusable blotting device requires less buffer and blotting paper, and refill packs are available
- Compact - has a small footprint and is stackable for multiple, simultaneous transfers
- Efficient - no heavy weights are required on the top of the transfer stack, eliminating the messy set-up of standard upward capillary transfers
- Easy to use - simple to set up and works without power or vacuum
The TurboBlotter System is a rapid downward blotting device for high-resolution transfer of DNA and RNA. It provides high-resolution alkaline transfers of DNA in less than one hour, and performs neutral (SSC) transfers of DNA and RNA within three hours. Larger device is stackable for multiple, simultaneous transfers. Reusable and easy to assemble, refills use Whatman Nytran SPC membrane and Whatman 3MM Chr and GB003 or GB005 blotting papers for top performance. The conventional Northern/Southern transfer stack has been turned upside down in order to take advantage of gravity. No heavy weights are required on the top of the transfer stack, eliminating the messy set-up of standard upward capillary transfers.
* Each TurboBlotter™ kit includes: Transfer device, 5 sheets of membrane, 40 sheets of 3MM Chr, 100 sheets of GB004, and 5 wicks of 3 MM Chr.
** TurboBlotter™ refills include: 5 sheets of membrane, 40 sheets of 3MM Chr, 100 sheets of GB004 and 5 wicks of 3 MM Chr.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Related Information
Certificates
Search by Lot Number28152-142_A22118265
Conformity declaration
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements