About this item
These stationary lounges are specially designed to accommodate space restrictions while assuring patient comfort and technician safety. The lounges can conveniently store 170 L (6cu.ft.) of supplies, easily move into the Trendelenburg position, and accommodate left or right draws from either side of the lounge.
- Linear actuator motor provides lasting and reliable functioning
- Five easy-to-replace seamless cushions are included, all of which reduce exposure to blood-borne pathogens
- The fabric contains an agent to protect against bacterial and fungal microorganisms
- Low entry and exit height for user convenience
Choice of 50.8 cm (20") or 68.5 cm (24") widths. Mobile coach configurations are available.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- CA80087-306
- CA80087-304
- CA80087-296
- CA80087-298
- CA80087-302
- CA80087-300
- Model:
- CA80087-306
- CA80087-304
- CA80087-296
- CA80087-298
- CA80087-302
- CA80087-300
- Weight capacity:
- 650 lbs.
- 650 lbs.
- 650 lbs.
- 650 lbs.
- 650 lbs.
- 650 lbs.