Sparging Relief Valve, Upchurch Scientific®
Catalog #: 53500-818
Supplier: Upchurch Scientific
Specifications
- Description:Sparging Relief Valve
- Cat. no.:53500-818
- Supplier No.:P-753
Specifications
About this item
The 3/16" ruby ball will quickly open and depressurize the reservoir before the mobile phase can rise up the tubing.
Customers Who Bought This Also Bought