Three-Way Valve, Model 7030, Rheodyne®
Catalog #: 50807-124
Supplier: Rheodyne
Specifications
- Description:Three-Way Valve, Model 7030
- Cat. no.:50807-124
- Supplier No.:7030
Specifications
About this item
Two-position six-port stainless steel valve with a double three-way switching pattern
Accepts 1/16" fittings.
Customers Who Bought This Also Bought