Synthware Distillation Receiving Sets, Vacuum Adapter, Kemtech America
Supplier: Kemtech America
Certificates
About this item
Designed without a hose connection for use with a distillation apparatus (see 60002-116 series).
- Heavy wall borosilicate construction
- Ground glass joint
- Extended drip tip
This distillation receiving set comes with a four leg cow receiver.
Ordering information: Set components are sold separately.
Customers Who Bought This Also Bought