Nunc® Replication Systems, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Designed for use with 96-well and 384-well plates (see 62409-602 series) and OmniTrays (see 62409-600 series).
- Pin replicators are ideal for transferring small volumes of inoculum from MicroWell™ plates to other plates or solid supports
- Replicator pins are durable stainless steel and are precisely aligned to ensure long-term performance even after repeated flame sterilization
Biodyne® B nylon membranes (62409-614) are precut to fit into the OmniTray, for manual or automated arraying directly on a solid support.
MicroWell™ Plate Copier (62409-610) and OmniTray Copier (62409-612) precisely register the replicator pins to agar, hybridization membranes, or the wells of MicroWell plates. Copiers are machined from polypropylene and are resistant to solvents and bleach.
Customers Who Bought This Also Bought