Crews® Stryke® Safety Goggles, MCR Safety
Supplier: MCR Safety
Certificates
About this item
Goggles offer reliable protection from chemical splash while providing a secure, ventilated fit.
- Stylish, smoke-tinted plastic frame
- Clear polycarbonate lens with Duramass® scratch-resistant coating
- Exclusive Duramass AF4® anti-fog coating
- Rubber strap adjusts easily
Goggles offer reliable protection from chemical splash while providing a secure, ventilated fit.
Certifications: Goggles meet or exceed ANSI Z87.1 safety standards.