Crews® Verdict® Protective Goggles, MCR Safety
Supplier: Avantor
Certificates
About this item
These safety goggles feature indirect ventilation for protection against chemical splash.
- Transparent protective PVC body permits a comfortable fit over prescription eyewear while protecting against impact
- Supplied with an elastic headband
- Lined goggles feature a soft foam lining that cushions the face and improves seal
- Can be worn with most half-mask respirators
These safety goggles feature indirect ventilation for protection against chemical splash.
Scratch-resistant polycarbonate lenses provide 99.9% UV protection and unobstructed panoramic visibility.
Certifications: Meet or exceed ANSI Z87.1+, CSA A94.3, and AS/NZS 1337-1992 standards.
Ordering information: All goggles feature an elastic headband except goggles 80081-795, which have an adjustable neoprene headband.