ContactUsLinkComponent
Sight Savers® Premoistened Lens Cleaning Tissues, Bausch & Lomb®
Sight Savers® Premoistened Lens Cleaning Tissues, Bausch & Lomb®
Catalog #: 52848-078
Supplier:  Bausch & Lomb
SIGHT SAVERS* Pre-Moistened Lens Cleaning Tissues
Sight Savers® Premoistened Lens Cleaning Tissues, Bausch & Lomb®
Catalog #: 52848-078
Supplier:  Bausch & Lomb
Specifications
  • Description:
    Premoistened Lens Cleaning Tissues
  • Cat. no.:
    52848-078
  • Supplier No.:
    8574GM
Specifications
About this item
These nonsilicone, antistatic tissues are individually packaged and treated with special surfactants to clean all glass and plastic lenses.
Perfect for computer screens, mirrors, safety glasses, and face shields. Unique package design allows easy dispensing of large 12.7×20.3 cm (5×8") folded, premoistened tissues. Cardboard dispenser box measures 22.9×13.7×6 cm (9×5³/₈×2³/₈").
Customers Who Bought This Also Bought