About this item
Suitable for high-performance separation of acidic, basic, nonpolar, and metal chelating compounds.
Chromolith® column speed comes from a highly porous monolithic silica gel rod clad to the column shell, which allows for high flow rates over a very large surface area. Superior, reproducible separations occur in less time than with packed-particle columns.
CapRod® capillary columns can achieve separations at 1 to 3 µL/min., rather than 200 to 400nL/min. for conventional media on a standard nano-LC capillary. FastGradient columns offer ultra-high performance combined with low operating pressure, and are ideal for applications in pharmaceutical, chemical, food, and academic labs. Flash and SpeedROD columns are ideal for high-speed separations of simple mixtures. Ideal for use in research laboratories, especially those specializing in combinatorial chemistry.
Performance columns provide rapid, high-quality separation of complex mixtures. Performance validation kit 48219-742 is ideal for use in quality control or validation laboratories. Kit contains three HPLC columns, each derived from a different batch in order to compare the batch-to-batch reproducibility and quality.
Guard cartridges are suitable for reversed-phase chromatography because they are all chemically modified in terms of carrying hydrophobic n-octadecyl groups on the surface of the monolithic silica rod. Guard cartridge kits consist of three pieces of guard cartridges, a cartridge holder, and a wrench. The column coupler features two 10-32 male connectors for female inlets of column.
Ideal as a first line routine column in the laboratory.
Specifications
- Catalog No:
- CA1.52002.0001
- CA1.52015.0001
- CA1.52014.0001
- CA48219-490
- CA48219-738
- CA48219-736
- CA48219-878
- CA97025-818
- CA82031-440
- CA48219-742
- CA48219-740
- CA48219-468
- CA48219-880
- CA97025-820
- CA97007-930
- Endcapped:
- Yes
- Yes