Uvex Classic™ 9305 Safety Goggles, Honeywell Safety
Supplier: Avantor
Certificates
About this item
Goggles are designed with closed vents for excellent protection.
Lenses are optically correct molded polycarbonate coated with Uvextreme® AF coating to provide antifog, anti-UV, anti-scratch and antistatic properties. Goggles are designed with closed vents for excellent protection.
Goggles are designed with closed vents for excellent protection.
Lenses are optically correct molded polycarbonate coated with Uvextreme® AF coating to provide antifog, anti-UV, anti-scratch and antistatic properties. Offer excellent splash protection. Soft, extra-wide frames fit snugly and comfortably around face, even over glasses. The wide neoprene strap holds goggles securely in place.
Certifications: Meet ANSI Z87.1 and CSA Z94.3 standards.
Customers Who Bought This Also Bought