Specifications
- Description:VersaQuat™ disinfectant and cleaner
- Packaging:Double bagged
- Volume:4 gal./512 oz.
- Packing Type:Trigger spray bottle
Specifications
About this item
VersaQuat™ is an EPA-registered, RTU (ready-to-use) one-step disinfectant and cleaner which utilizes well-established and accepted quaternary ammonium chemistry with acceptable residue and safety profiles. Made in the USA with no preservatives or fragrances, Berkshire’s VersaQuat™ is a powerful and safer disinfectant alternative for cleaning and disinfecting needs.
- Kills common microbiological contaminations on surfaces
- Can be used in a variety of applications
- Ideal for large surface cleaning
- No strong odor provides good user acceptability
- No special disposal is required
- Facilitates transfer disinfection into a cleanroom
- No mixing required
With broad-spectrum, best-in-class efficacy claims (30 kill claims), VersaQuat™ delivers consistent quality and intuitive usage. Recommended for use as a standard disinfectant as part of a disinfectant rotation program, VersaQuat™ is ideal for use in laboratories, hospitals, home healthcare settings, and veterinary clinics. It can also be used in offices, schools, gyms, and homes. Quick, effective, and convenient, VersaQuat™ is not abrasive and is compatible with various hard, non-porous surfaces.