About this item
Space-saving multi-action platform rotator/rocker produces six different mixing motions: rotating, rotating/rocking, rotating/rolling, rolling/rocking, and rocking/shaking, as well as a combination of all of these.
- Multi-action platform
- Six mixing motions
- Magnetized platform
The optional 3-D orbital shaker attachment transforms the unit into an orbital shaker
Clip plates hold similar or different tube sizes for various mixing motions, which can be achieved by varying the orientation of the clip plates on the magnetized platform. Ideal for use in cold rooms and low-temperature incubators. Complete and even soaking of membrane-based blot assays/washings. Easily cleaned and decontaminated. Rotator/rocker maintains set speed at temperatures from 0 to 38°C (32 to 100°F). Expansion kits are available for increased capacity and different containers.
The easily-removable, double-sided, stainless steel, magnetized platform converts from rotating to rocking with a simple mechanical adjustment. Platform can accommodate up to two 15.2x15.2cm (6x6") plastic bags per side, a 30.5x30.5cm (12x12") nonskid metal rocker tray for conventional rocker/shaker operations, or up to four metal clip plates per side.
Specifications
- Catalog No:
- 14003-580
- 14222-144
- 15001-012
- 15001-014
- 89401-428
- 58815-176
- 15001-010
- 14222-140
- 89426-872
- 15001-008
- 76808-692
- 15001-006
- 15001-004
- 15001-002
- 14003-576
- 490015-482
- 10753-556
- Rocking angle:
- ±10° From Horizontal
- Unit dimensions:
- 27H x 51W x 25D cm (101/2 x 20 x 97/8") without tray
- Platform dimensions:
- 20.3W x 34.3L cm (8 x 131/2")
- Weight capacity:
- 4.5 kg (10 lbs.)
- Weight:
- 11.8 kg (26 lbs.)
- Max. capacity:
- 96 Tubes, 10–13 mm Diameter
48 Tubes, 15–17 mm Diameter
24 Tubes, 28–30 mm Diameter
4 Plastic Bags, 15.2 x 15.2 cm (6 x 6")
8 Metal Clip Plates