J.T.Baker® BAKERBOND®, Narrow-Pore Columns
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Non-Regulated, With Information
About this item
For analytical and preparative HPLC.
All columns incorporate proprietary bonding chemistry on 5 µm narrow-pore spheroidal silica gel particles with an average pore diameter of 120 Å. These packings are chemically stable from pH 2 to 8.5. They are not damaged by the use of cleaning and antimicrobial agents such as sodium azide, dilute aqueous solutions of trifluoroacetic acid and acetic acid, organic solvents such as methanol and acetonitrile, and concentrated aqueous solutions of salts and urea. Columns have an operating pressure of 1000 to 1500 psi.
Related Information
Certificates
Search by Lot Number7098 Product Specifications
Additional Product Details
Avantor JT Baker Extraction of Explosives and Metabolites from Water Application Note
Product Literature
spec_JT7098-0
Product specification
Customers Who Bought This Also Bought