Synthware Connecting Adapters, Kemtech America
Catalog #: 60000-322
Supplier: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- ST Joint:24/40
- Cat. no.:60000-322
- Supplier No.:A192424
Specifications
About this item
Converts standard vessels to airless applications.
- [ST] joint
- PTFE stopcock
Features a 2 mm PTFE stopcock.
Customers Who Bought This Also Bought