VWR® Critical Swab® Swab, Cone-Shaped Reticulated Foam Head
Supplier: VWR International
CE Compliant
Certificates
About this item
Swab features reticulated foam and a blue glass-filled polypropylene shaft for extra rigidity.
- Reticulated foam
- Polypropylene shaft
- Extra rigid
For use in critical environments.
Related Information
Certificates
Search by Lot NumberPuritan Controlled Environments Industrial Products
Product Literature
Customers Who Bought This Also Bought