Cleanroom Labels Autoclave, TX515A
Catalog #: TW-TX515A
Supplier: Texwipe
New Product
Specifications
- Color:White
- Description:Autoclavable labels, Teslin®
- Packaging:500 labels/roll
- Dimensions:38 mm
Specifications
About this item
These autoclavable labels are made of a Teslin® material designed for label printing and can be used with most conventional inks.
- Matte finish
- Permanent acrylic adhesive to provide excellent adhesion
- Prevent the removal of written data during the autoclave process
Ideal for applications requiring label and label data to maintain integrity through autoclave process.
Customers Who Bought This Also Bought