Order Entry
ContactUsLinkComponent
Urine Quad Plate
Urine Quad Plate
Catalog #: CA77497-256
New Product
CAS Number:  
Urine Quad Plate
Catalog #: CA77497-256
New Product
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Packaging:
    10 per package
  • Description:
    Urine quad plate (Eosine methylene blue (EMB) agar, levine/xylose lysine deoxycholate (XLD) agar, modified/simmons citrate/blood agar, 5%)
  • Size:
    1.5×10 cm
  • Plate Size:
    1.5×10 cm
  • Application:
    Clinical industry

Specifications

About this item

EMB Agar formulations are recommended for use as selective and differential media for the isolation of Gram-negative bacilli (including coliform organisms and enteric pathogens) from clinical and nonclinical specimens.

  • Optically clear polystyrene
  • Heavy-Duty
  • Stacking rings
  • Breathable packaging

XLD Agar is recommended for use as a selective and differential medium for the isolation of Gram-negative enteric pathogens from fecal specimens or from food and other samples.

Citrate Agar is recommended for use in the differentiation of Gram-negative enteric bacilli based on citrate utilization.

Blood Agar products are recommended for use as general purpose growth media for the isolation, cultivation, and differentiation of a wide variety of microorganisms.

Customers Who Bought This Also Bought