Rtx®-1 GC Columns, Restek
Supplier: Restek
CE Compliant
Certificates
About this item
These general-purpose columns are designed for solvent impurities, PCB congeners, simulated distillation, drugs of abuse, gases, natural gas odorants, sulfur compounds, essential oils, hydrocarbons, semi-volatiles, pesticides, and oxygenates.
- Low bleed levels
- Longer column lifetime
- Short conditioning times
Columns exhibit long lifetimes and very low bleed at high operating temperatures. A proprietary synthesis process eliminates residual catalysts that could cause degradation and increase bleed. Phase composition: crossbonded 100% polydimethylpolysiloxane. Equivalent to USP G1, G2, and G38 phases.
Specifications
- Catalog No:
- RK10144
- RK10142
- 10852-072
- 10850-730
- 10058-080
- 10058-076
- 10058-078
- 10850-650
- RK10128
- RK10137
- 10852-092
- 10850-264
- 10850-266
- 10850-268
- 10850-270
- RK10105
- RK10107
- RK10106
- RK10111
- RK10109
- 10850-822
- 10850-824
- 10058-094
- 10058-096
- RK10112
- 75851-582
- 10058-090
- RK10120
- 10058-092
- 10803-454
- RK10108
- RK10123
- RK10135
- RK10121
- RK10124
- RK10139
- RK10138
- RK101411
- 10850-606
- 10058-074
- 10850-610
- RK10143
- 10850-638
- RK10140
- 10059-012
- RK10136
- RK10129
- RK10127
- RK10126
- 10850-628
- RK10125
- RK10122
- Description:
- Rtx-1
- Rtx-1
- Rtx-1
- Rtx-1 with 5 m guard column
- Rtx-1
- Rtx-1
- Rtx-1
- Rtx-1 with 5 m guard column
- Rtx-1
- Rtx-1
- Rtx-1
- Rtx-1
- Rtx-1 with 5 m guard column
- Rtx-1 with 5 m guard column
- Rtx-1 with 5 m guard column
- Rtx-1
- Rtx-1
- Rtx-1
- Rtx-1
- Rtx-1
- Rtx-1
- Rtx-1
- Rtx-1
- Rtx-1
- Rtx-1
- Rtx-1 with 5 m guard column
- Rtx-1
- Rtx-1
- Rtx-1
- Rtx-1
- Rtx-1
- Rtx-1
- Rtx-1
- Rtx-1
- Rtx-1
- Rtx-1
- Rtx-1
- Rtx-1
- Rtx-1
- Rtx-1
- Rtx-1 with 10 m guard column
- Rtx-1
- Rtx-1 with 5 m guard column
- Rtx-1
- Rtx-1
- Rtx-1
- Rtx-1
- Rtx-1
- Rtx-1
- Rtx-1
- Rtx-1
- Rtx-1