ContactUsLinkComponent
SP Bel-Art Poxygrid® 96-Place Test Tube Rack, Bel-Art Products, a part of SP
SP Bel-Art Poxygrid® 96-Place Test Tube Rack, Bel-Art Products, a part of SP
Catalog #: 47729-698
Poxygrid* 96-Place Test Tube Rack
SP Bel-Art Poxygrid® 96-Place Test Tube Rack, Bel-Art Products, a part of SP
Catalog #: 47729-698
Specifications
  • Cat. no.:
    47729-698
  • L×W×H:
    24.2 x 17 x 6.4 cm (91/2 x 611/16 x 21/2")
  • Supplier No.:
    187650001
Specifications
About this item
Rack emulates the 96-place format used in microplates, deep well plates, and other tools configured for drug research and robotic applications.
  • Chemical- and corrosion-resistant
  • Epoxy-coated steel wire construction
  • Unaffected by organic solvents and weak acids and alkalis
  • Autoclavable
Flow-through design allows rapid temperature change for samples suspended in a water bath. Holds ninety-six 13 mL -16 mL tubes in an 8 x 12 configuration. Chemical and corrosion-resistant steel wire is coated with heat-cured epoxy and are unaffected by organic solvents, salts, weak acids, and weak alkalis. Steam autoclavable at 121°C (250°F).
Customers Who Bought This Also Bought