Accessories for 360 ml Stainless Steel Container for Waring® Blenders, Eberbach
Supplier: Eberbach
Certificates
Certificates
About this item
Customers Who Bought This Also Bought
Acetonitrile ≥99.9%, OmniSolv® LC-MS for LC-MS, Supelco®
Supplier: MilliporeSigma
$1,313.27 to $5,149.78
Masterflex® Luer Stopcock Fittings, Polycarbonate, Avantor®
Supplier: VWR International
$64.62 to $66.68
Blood Agar Plate, 5% Sheep Blood in Tryptic Soy Agar (TSA) Base, 15×100 mm plate
Supplier: HARDY DIAGNOSTICS CA
$42.99
Clarity Urocheck Urinalysis Strips, Clarity Diagnostics
Supplier: Clarity Diagnostics
$31.63 to $756.61