About this item
These syringes are compatible with Cavro XL syringe pump, Centris, X-CMP and Tecan Fluent instruments.
- Front fitting: ¹/₄"-28 UNF
- Glass barrel: Borosilicate 3.3
- Precision: <±1% of the volume
- Stroke: 30 mm
PTFE-seal: Chemically resistant; heavy duty syringes.
PTFE-seal with silicone: Specially designed for handling of strong buffers and other aqueous solutions. The modified PTFE-seals guaranteed, extended lifetime under heavy duty conditions.
HDPE-seal: Specially designed for handling of strong buffers and other aqueous solutions. The special PE-seals run smoother than conventional seals. Good resistance to buffers and aqueous solutions, long lifetime.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.