Synthware Drying Chambers, Abderhalden, Kemtech America
Catalog #: 60002-154
Supplier: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- Cat. no.:60002-154
- Bottom [ST] Joint Size:24/40
- Top ST Joint Size:[ST] 24/40
- Supplier No.:D222434
Specifications
About this item
Drying samples for elemental analyzis.
- Abderhalden style
- Heavy wall borosilicate construction
- Ground glass joint
Sample is placed in a vial which is then placed into drying chamber. Drying chamber [ST] joint size: 34/45.
Customers Who Bought This Also Bought