About this item
These syringes are used for Zinsser analytic lissy/sophas/speedy/calli/moss, rosys plato and lucy, qiagen biorobot 9600, digene* rapid capture system, unitech chemwell* 2910.
- Front fitting: Teflon Luer Lock (TLL)
- Plunger top: Hole Ø 4.8 with screw
- Stroke: 60 mm
- Glass barrel: Borosilicate 3.3
- Precision: <±1% of the volume
PTFE-seal: Chemically resistant; heavy duty syringes.
PTFE-seal with silicone: Specially designed for handling of strong buffers and other aqueous solutions. The modified PTFE-seals guaranteed, extended lifetime under heavy duty conditions.
HDPE-seal: Long lifetime; good resistance to strong buffers and other aqueous solutions.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.