Synthware Condenser for Rotary Evaporators, Kemtech America
Catalog #: 60001-176
Supplier: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- Description:Rotary Evaporator Condenser, Diagonal-Style
- Cat. no.:60001-176
- Supplier No.:C910200
Specifications
About this item
Double-coil condenser for use with rotary evaporators for cooling with tap water in a diagonal position.
- Heavy wall Borosilicate construction
- 19/38 Ground GlassTop joint
- Buchi style condenser
- 35/20 Spherical joint
With a [ST]19/38 top valve and 35/20 spherical ball joint.
Customers Who Bought This Also Bought