Order Entry
ContactUsLinkComponent
Chemdye® PRO1 RT Hygiene Monitoring System
Chemdye® PRO1 RT Hygiene Monitoring System
Catalog #: 77506-910
Supplier:  TERRAGENE
New Product
PROTEIN DETECTION SEMI-QUAN
Chemdye® PRO1 RT Hygiene Monitoring System
Catalog #: 77506-910
Supplier:  TERRAGENE
New Product

Specifications

  • Description:
    PRO1 RT hygiene monitoring semi quantative protein detection system

Specifications

About this item

Semi quantitave protein detection system. Designed to detect and quantify proteins with high sensitivity on surfaces, solution and difficult-to-access areas after the cleaning process.

  • Semiquantitave results comparing with color chart
  • Pen diameter: 12.4 mm; Pen length: 161 mm; Swab length: 111.9 mm
  • Limit of detection: 25 μg or 0.5 mg/ml of BSA
  • Shelf-life: 2 years when stored refrigerated (2 to 8 °C) or 6 months without refrigeration (up to 30 °C)

The system consists of a reactive solution and a high absorption swab, which allows the collection of samples from different types of surfaces with the same efficiency.

Analysis: Incubate at room temperature (20 to 30 °C) for 10 minutes and interpret visually by using the color reference guide available inside the product’s box.

Shelf-life: 2 years when stored refrigerated (2 to 8 °C) or 6 months without refrigeration (up to 30 °C). 1 month if not refrigerated storage reduces de expiration date by 4 months.

Storage conditions:
T = 2 to 30 °C, RH = 30 to 80%, keep in a dark place. Store vertically as indicated in the package.

Certifications: ISO 13485.
Packaging: 100 per package.
Customers Who Bought This Also Bought