Synthware Four Finger Clamp, Kemtech America
Catalog #: 60005-030
Supplier: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- Description:Synthware four finger clamp
- Supplier No.:W170004
Specifications
About this item
Four finger clamp is constructed of die-cast alloy, with a nickel/chrome plated finish.
- Cork lined fingers
- Adjustable
- Chromed plated
- Plastic wing nut style tension screw
Clamps feature cork lined interlocking jaws. They accept articles up to 80 mm in diameter.
Customers Who Bought This Also Bought