VWR® Talon® Thermometer Extension Clamp
Catalog #: 21573-071
Supplier: VWR International
Product available on GSA Advantage®
Specifications
- Description:Thermometer Extension Clamp
- Cat. no.:21573-071
Specifications
About this item
Hold thermometers and other glass apparatus firmly with this lightweight, safety-adjust clamp.
- Lightweight
- Safely adjustable
- Nickel-plated phosphor bronze jaws
Holds 6.4–12.7 mm (1/4–1/2") diameter glass tubing/thermometers up to 178 mm (7") from support rod. Tightening wing nut applies tension to the nickel-plated phosphor bronze jaws. Overall length: 210 mm (81/4"). If attaching to rods, a 90° clamp holder is required.
Customers Who Bought This Also Bought