ContactUsLinkComponent
Cocoon Environmental Enrichment Bedding, Ketchum
Cocoon Environmental Enrichment Bedding, Ketchum
Catalog # 95043-336
undefined
Cocoon Environmental Enrichment Bedding, Ketchum
Catalog # 95043-336
Supplier Number:  XKA 2600 087
Specifications
  • Description:
    Cocoon Environmental Enrichment Bedding
  • Cat. no.:
    95043-336
  • Supplier No.:
    XKA 2600 087
Specifications
About this item
Designed to benefit the health and contentment of rodents used for research, the Cocoon offers animals the oppurtunity to engage in social activity and fulfill nesting instincts
The small, lightweight, cylindrical form of the Cocoon makes it easily moveable, increasing the interaction between the animal, the enrichment product, and the environment. The cylinder is constructed of short cotton fibers that are safe for the animal and its offspring. Once torn, the Cocoon can be modified to create a vaporous cloud which can be used to create a nest. Cocoon is whitened without chlorine. Packaged 6,000 cylinders per pack.