To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Clonality:Monoclonal
- Clone:14H4
- Gene ID:8829
- Host:Mouse
- Isotype:IgG 2b/k
- Antigen symbol:CD304
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:100 µg
- Western blot:Yes
- Form:Purified
- Shipping temperature:Store undiluted between 2...8 °C
- Supplier No.:652302
Specifications
About this item
The membrane isoform of CD304 is a receptor that regulates angiogenesis and organogenesis in response to growth factors of the VEGF family. CD304 forms a complex with VEGFR2/KDR, resulting in increased cell migration and chemotaxis. In the nervous system, CD304 complexed with plexin mediates Sema-3A signaling and may be involved in axon guidance.
Type: Primary
Antigen: CD304
Clonality: Monoclonal
Clone: 14H4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human