Foreline Trap, Edwards
Catalog #: CAA133-05-000
Supplier: Edwards
Specifications
- Description:Foreline Trap
- Cat. no.:CAA133-05-000
- Supplier No.:A133-05-000
Specifications
About this item
For clean pumping systems to prevent back-migration of rotary pump oil vapor into the system
Note: To minimize vapor absorption during pump down, a valved bypass line is recommended.
It is ideal for two-stage pumps operating below 1mbar. Trap is filled with activated alumina, which has excellent oil vapor trapping properties. For models RV3, RV5, RV8, and R12.
Customers Who Bought This Also Bought